
6,6'-Dimethyl-2,2'-bipyridyl 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4411-80-7
:
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00059776
Boiling Pt:286°C
:184.2371 g/mol
EINECS:224-566-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to yellow crystalline powder and or chunks
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Melting point:86°C to 96°C
- Water:≤1 %
Specification Test Results
Frequently Bought Together