2-Ethylhexyl salicylate
New Product
:118-60-5
:
:C₁₅H₂₂O₃
:MFCD00053300
:250.34 g/mol
:190°C (21mmHg)
:1.02
:162°C
:Ambient
:204-263-4
- Appearance:clear colorless to light yellow liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Refractive index:1.5010 to 1.5030 (20°C, 589 nm)
Frequently Bought Together