Wright's stain for microscopical staining, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:68988-92-1
:51938-72-8
MDL Number:MFCD00082143
Density:0.8 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ400
EINECS:273-541-5
:GHS07
Storage Temperature:Ambient
:1
Frequently Bought Together