Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium silicate
Magnesium silicate for chromatography
Magnesium silicate for chromatography
Magnesium silicate for chromatography
Catalog # SUPL20281
Supplier:
Merck
CAS Number:
Expand image
Magnesium silicate for chromatography
Catalog # SUPL20281
Supplier:
Merck
CAS Number:
Specifications
Pk:
1
Seq:
0001
Specifications
About this item
Product Details
:
:
100.38 g/mol
EINECS:
215-681-1
CAS Number:
1343-88-0
Formula:
MgO₃Si
MDL Number:
MFCD00078249
Product Details