

Dicyclohexylcarbodiimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:538-75-0
:
Formula:C₁₃H₂₂N₂
MDL Number:MFCD00011659
Merck Index:13,03123
Boiling Pt:148…152 °C (15 hPa)
Melting Pt:35…36 °C
Density:0.95 g/cm³ (40 °C)
Flash Pt:113 °C
EINECS:208-704-1
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:206.33 g/mol
:GHS06
:GHS05
Related Information
Certificates
Search by Lot NumberMERC8.02954.0250_TDS_EN ProductPage_802954
Product specification
msds_802954
SDS
Frequently Bought Together