4-Amino-3-hydroxy-1-naphthalenesulphonic acid, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:116-63-2
:
Formula:C₁₀H₉NO₄S
MDL Number:MFCD00004019
:239.25 g/mol
Merck Index:13,00453
Boiling Pt:466 °C (1013 hPa)
Melting Pt:∼285 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:204-147-3
Frequently Bought Together