Nitro blue tetrazolium chloride (NBT), Millipore®
Supplier: Merck Millipore (Oncogene)
Certificates
About this item
Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
CAS Number:298-83-9
:
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159
Molecular Weight:817.65 g/mol
Melting Pt:200
EINECS:206-067-4
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together