
Mupirocin
New Product
:12650-69-0
:
:C₂₆H₄₄O₉
:MFCD01711620
:500.629 g/mol
:GHS07
:Freezer
Frequently Bought Together
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
3-(Chloromethyl)benzoyl chloride 98%
: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Liquid for manometer with sloping tube
: VWR Chemicals