Aluminium chloride hexahydrate, EMPROVE® ESSENTIAL, meets analytical specification of BP, Ph. Eur., USP
Supplier: MERCK PRODUCTION CHEMICALS
New Product
Certificates
About this item
CAS Number:7784-13-6
:
Formula:AlCl₃·6H₂O
MDL Number:MFCD00149134
:241.43 g/mol
Melting Pt:100 °C
UN:3260
ADR:8,II
:GHS05
:GHS08
Storage Temperature:Room Temperature
Frequently Bought Together