2,2'-Methylenebis(6-tert-butyl-p-cresol) 99+%
Supplier: Apollo Scientific
New Product
Certificates
About this item
CAS Number:119-47-1
:
Formula:C₂₃H₃₂O₂
MDL Number:MFCD00043641
:340.51 g/mol
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Bottles, narrow neck, round, HDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck