Order Entry
Italy
ContactUsLinkComponent
GANT58 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
GANT58 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 475864-5
CAS Number:  
GANT58 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 475864-5
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :392.4756 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:64048-12-0
Formula:C₂₄H₁₆N₄S
MDL Number:MFCD01103196