L-779450 ≥97% (by HPLC), Calbiochem®, Sigma-Aldrich®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:303727-31-3
:
Formula:C₂₀H₁₄ClN₃O
MDL Number:MFCD12828750
:347.80 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
Frequently Bought Together