ABT-491 hydrochloride ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 511071-2
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:2 mg
- Seq:2145
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
:479.50 g/mol (free base basis)
CAS Number:189689-94-9
Formula:C₂₈H₂₂FN₅O₂·xHCl
MDL Number:MFCD00945301