Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trimethylamine
Trimethylamine ∼8% in DMF
Trimethylamine ∼8% in DMF
Trimethylamine ∼8% in DMF
Catalog # TCIAT3847-100G
Supplier:
TCI
CAS Number:
Expand image
Trimethylamine ∼8% in DMF
Catalog # TCIAT3847-100G
Supplier:
TCI
CAS Number:
Specifications
Pk:
100 g
Specifications
About this item
Product Details
:
:
59.11 g/mol
UN:
2920
CAS Number:
75-50-3
Formula:
C₃H₉N
MDL Number:
MFCD00008327
Product Details