Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Propene
Propene 1-2% in toluene
Propene 1-2% in toluene
Catalog # TCIAP2815-100ML
Supplier:
TCI
CAS Number:
Expand image
Propene 1-2% in toluene
Catalog # TCIAP2815-100ML
Supplier:
TCI
CAS Number:
Specifications
Pk:
100 mL
Specifications
About this item
Product Details
:
:
42.08
UN:
1993
CAS Number:
115-07-1
Formula:
C₃H₆
Product Details