Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium di-tert-butyl phosphate
Potassium di-tert-butyl phosphate
Potassium di-tert-butyl phosphate
Potassium di-tert-butyl phosphate
Catalog # TCIAP2797-1G
Supplier:
TCI
CAS Number:
Expand image
Potassium di-tert-butyl phosphate
Catalog # TCIAP2797-1G
Supplier:
TCI
CAS Number:
Specifications
Pk:
1 g
Specifications
About this item
Product Details
:
:
248.3 g/mol
CAS Number:
33494-80-3
Formula:
C₈H₁₈KO₄P
MDL Number:
MFCD03840344
Product Details