Order Entry
Italy
ContactUsLinkComponent
 
(trans,trans)-4'-Propyl[1,1'-bicyclohexyl]-4-methanol
Catalog # TCIAP2432-5G
Supplier:  TCI
CAS Number:  
(trans,trans)-4'-Propyl[1,1'-bicyclohexyl]-4-methanol
Catalog # TCIAP2432-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

 :
 :238.41 g/mol
CAS Number:82562-85-4
Formula:C₁₆H₃₀O
MDL Number:MFCD14636523