2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione
Catalog # TCIAN1054-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
Specifications
About this item
:
:329.231 g/mol
CAS Number:104206-65-7
Formula:C₁₄H₁₀F₃NO₅
MDL Number:MFCD01752192