Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,8-Didodecyloxy-13-methoxytriptycene
1,8-Didodecyloxy-13-methoxytriptycene
1,8-Didodecyloxy-13-methoxytriptycene
1,8-Didodecyloxy-13-methoxytriptycene
Catalog # TCIAD5881-100MG
Supplier:
TCI
CAS Number:
Expand image
1,8-Didodecyloxy-13-methoxytriptycene
Catalog # TCIAD5881-100MG
Supplier:
TCI
CAS Number:
Specifications
Pk:
100 mg
Specifications
About this item
Product Details
:
CAS Number:
1798293-53-4
Product Details