Order Entry
Italy
ContactUsLinkComponent
(2-(3,6-Dimethoxy-9H-carbazol-9-yl)ethyl)phosphonic acid
(2-(3,6-Dimethoxy-9H-carbazol-9-yl)ethyl)phosphonic acid
Catalog # TCIAD5798-500MG
Supplier:  TCI
CAS Number:  
(2-(3,6-Dimethoxy-9H-carbazol-9-yl)ethyl)phosphonic acid
Catalog # TCIAD5798-500MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mg

Specifications

About this item

 :
 :335.29 g/mol
CAS Number:2377770-18-6
Formula:C₁₆H₁₈NO₅P