Showa 234 Cut Resistant Gloves, Foam Nitrile Dipped
Supplier: Showa
Certificates
About this item
Designed for use in 'knife hand' applications in food processing.
- Cut-resistant and abrasion-resistant
- Flexible and breathable
- Lightweight and increased dexterity
- Machine washable
- Seamless knit
- Heat-resistant and hydrocarbon-resistant
- Form-fitting
This flexible cut protection glove combines foam nitrile with a HPPE reinforced liner to provide excellent cut resistance performance. A foam nitrile coating protects against oils, hydrocarbons, grease, and abrasions, while offering excellent grip in wet and dry conditions. Cooling properties of HPPE and a breathable design reduce perspiration to keep hands dry.
Certifications: Cat II. EN 388:2016+A1:2018, 4X42D. EN 407:2020, X1XXXX. Food contact approved.
Packaging: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pair per case).
Specifications
- Catalog No:
- BEST234-9/XL
- BEST234-7/M
- BEST234-8/L
- BEST234-10/XXL
- BEST234-6/S
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile