Order Entry
Italy
ContactUsLinkComponent
Cole-Parmer® Cleanroom Hose Barb Fitting
Cole-Parmer® Cleanroom Hose Barb Fitting
Catalog # STUA50108-36
Supplier:  Cole Parmer
New Product
Cole-Parmer® Cleanroom Hose Barb Fitting
Catalog # STUA50108-36
Supplier:  Cole Parmer
New Product
Specifications
  • Barb size/tubing int.Ø (port 1):
    ¹/₈"
  • Barb size/tubing int.Ø (port 2):
    ¹/₈"
  • Connection Type:
    Hose barb union
  • Flow pattern:
    Straight
  • Max. temperature:
    66 °C (150 °F)
  • Colour:
    Natural
  • Description:
    Hose barb fitting, straight union, animal-derivative free polypropylene, cleanroom packed, ¼" int.Ø
  • Min. temperature:
    –23 °C (–9 °F)
  • Sterilisation:
    Gamma irradiation
  • Pk:
    10
  • Fitting material:
    Polypropylene
Specifications
About this item
This is used in cleanroom to minimise contamination by pollutants.
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly 'grips' the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0,5 µm or larger per cubic foot.
Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidising agents.