
Ammonium iron (III) citrate, green
Catalog # HIMEGRM476-500G
Supplier: HIMEDIA
New Product
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
EINECS:214-686-6
:GHS07
Storage Temperature:Room Temperature
CAS Number:1185-57-5
Formula:C₆H₅+4y)Fe(x)N(y)O₇
MDL Number:MFCD00013099