Buccutite™ Rapid PerCP Antibody Labelling Kit
Catalog # AATB5407
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:2
- Includes:2 labellings
Specifications
About this item
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PerCP antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PerCP, APC, PE and iFluor® tandem dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped with blue ice.
Caution: Research use only.