Order Entry
Italy
ContactUsLinkComponent
Cell Meter™ Glucose Uptake Imaging Kit
Cell Meter™ Glucose Uptake Imaging Kit
Codice catalogo AATB23501
Supplier:  AAT Bioquest
New Product
CAS Number:  
Cell Meter™ Glucose Uptake Imaging Kit
Codice catalogo AATB23501
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Tests per kit:
    100 tests
Specifications
About this item
Red Fluorescence. The Cell Meter™ Glucose Uptake Imaging Kit provides a simple and direct method for quantifying glucose uptake across diverse cellular contexts.
  • Ex/Em = 651/670 nm
  • Quantification via both fluorescence microscopy and flow cytometry techniques
  • Six-month shelf life
This kit employs Glutite™ Red 670, a cell-permeable fluorescent glucose tracer, enabling precise molecular sensing and bioimaging via GLUT transporters.
Delivery: Shipped at RT.
Caution: Research use only.