Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Supelco® TraceCERT® Organic Reference Standard, Spinetoram Solution

Spinetoram solution, 1 mL
Catalog # 43065-1ML
Supplier:   Merck
Supelco® TraceCERT® Organic Reference Standard, Spinetoram Solution
Catalog # 43065-1ML
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Spinetoram Solution

Spinetoram solution, 1 mL

CAS Number:  
Price
96,80 €
Log in for availability
1 * 1 mL
Product Details & Documents

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.

Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Spinetoram solution
  • Cat. No.:
    43065-1ML
  • M-Clarity quality level:
    MQ300
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • CAS No.:
    935545-74-7

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.