Supelco® Organic Reference Standard, HMDI
33490-2ML-R
: Merck
:
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:5124-30-1
- Description:HMDI
- Storage conditions:2...30 °C
- Hazard class description:H315 - H317 - H319 - H330 - H334 - H335
- Shipping conditions:STD
- MDL No.:MFCD00015405
- M-Clarity quality level:MQ100
- Synonym(s):4,4′-Methylene-bis(cyclohexyl isocyanate); 4,4′-Diisocyanato-methylenedicyclohexane; 4,4′-HMDI; Bis(4-isocyanatocyclohexyl)methane
- Formula:CH₂(C₆H₁₀NCO)₂
- Molecular weight:262,35
- Flash point:200 °C - closed cup
- GHS signal: GHS06, GHS08
: P260 - P264 - P280 - P302 + P352 - P304 + P340 + P310 - P305 + P351 + P338