Supelco® Organic Reference Standard, Cis,Cis-5,8-Tetradecandienoyl-L-Carnitine
19188-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:1469901-01-6
- Description:cis,cis-5,8-Tetradecandienoyl-ʟ-carnitine
- Storage conditions:–20 °C
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):(2R)-3-Carboxy-N,N,N-trimethyl-2-[[(5Z,8Z)-1-oxo-5,8-tetradecadien-1-yl]oxy]-1-propanaminium inner salt
- Molecular weight:367,52