Order Entry
Italy
ContactUsLinkComponent
Magicard™ CR80 Rewritable Cards
Magicard™ CR80 Rewritable Cards
Catalog # BRDY322005
Supplier:  Brady
New Product
undefined
Magicard™ CR80 Rewritable Cards
Catalog # BRDY322005
Supplier:  Brady
New Product

Specifications

  • Colour:
    White
  • Description:
    CR80 rewritable cards
  • Length:
    86 mm
  • Material:
    PVC
  • Seq:
    06
  • Pk:
    100
  • Width:
    54 mm

Specifications

About this item

Plain white PVC card (CR80).

  • Re-write a card mutliple times

Pre-approved for best quality printing on Magicard™ printers. Cards are compatible with Magicard™ 300, Magicard™ 600, Pronto 100, and Prima 8 printers.

Packaging: 100 pieces per pack.