Order Entry
Italy
ContactUsLinkComponent
Supelco® Cerilliant® Organic Reference Standard, Acylcarnitines Mix 3 (C8:1,C10:1,C12:1,C14:1,C14:2,C16:1,C18:1,C18:2) solution
Supelco® Cerilliant® Organic Reference Standard, Acylcarnitines Mix 3 (C8:1,C10:1,C12:1,C14:1,C14:2,C16:1,C18:1,C18:2) solution
Catalog # A-145-1ML
Supplier:  Merck
CAS Number:  
Supelco® Cerilliant® Organic Reference Standard, Acylcarnitines Mix 3 (C8:1,C10:1,C12:1,C14:1,C14:2,C16:1,C18:1,C18:2) solution
Catalog # A-145-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 - 200 µg/ml (Methanol:1% aqueous formic acid (90:10) (v/v))
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • Description:
    Acylcarnitines mix 3 (C8:1,C10:1,C12:1,C14:1,C14:2,C16:1,C18:1,C18:2) solution
  • Storage conditions:
    –70 °C
  • Shipping conditions:
    DICE
  • Flash point:
    9,7 °C - Closed cup - Not applicable
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item