Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, BDE No 37
Supelco® Organic Reference Standard, BDE No 37
Catalog # 91408-10MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, BDE No 37
Catalog # 91408-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    147217-81-0
  • Description:
    BDE No 37
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H318 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01940710
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,4,4′-TriBDE; 3,4,4′-Tribromodiphenyl ether
  • Melting point:
    48 - 52 °C
  • Molecular weight:
    406,90

Specifications

About this item

Caution: P273 - P280 - P305 + P351 + P338 - P391 - P501