Supelco® Organic Reference Standard, Hexanoyl-ʟ-carnitine
91388-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:22671-29-0
- Description:Hexanoyl-ʟ-carnitine
- Storage conditions:2…8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-1-propanaminium inner salt; (R)-Caproylcarnitine; n-Hexanoyl-ʟ-carnitine; C6-Carnitine
- Molecular weight:259,34