Supelco® Organic Reference Standard, Mineral oil standard mixture Type A and B for EN 14039 and ISO 16703
Catalog # 69246-10ML-F
Supplier: Merck
New Product
CAS Number:
Specifications
- Concentration:~8 mg/ml in heptane (exact content on the label)
- Pk:10 mL
- Standards:Analytical standard
- Description:Mineral oil standard mixture Type A and B for EN 14039 and ISO 16703
- Storage conditions:2...30 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ200
- Density:0,686 g/ml at 20 °C
- Flash point:–4 °C
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331