Anti-IgG Goat Polyclonal Antibody
Supplier: AAT BIOQUEST
New Product
Certificates
About this item
Anti-Human IgG (Fc specific) antiserum is produced in goat using purified human IgG, Fc fragment, as the immunogen.
- No reactivity is observed with the Fab fragment of human IgG, light chains, IgA, or IgM
- No interspecies cross reactivity with mouse or rat serum proteins
- Purified through affinity chromatography
The antibody preparation is specific for human IgG, Fc fragment when tested against purified human IgA, IgG (Fc and Fab fragments).
Caution: Research use only.
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Fc)
Reactivity: (Fc)
Specifications
- Catalog No:
- AATB50302
- AATB50301
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Control
- Control
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Goat
- Goat
- Isotype:
- IgG Fc
- IgG Fc
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- Refrigerated at 2...8 °C, minimise light exposure
- Refrigerated at 2...8 °C, minimise light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together