Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nuclear Blue™ LCS1
Nuclear Blue™ LCS1
Nuclear Blue™ LCS1
AATB17559
:
AAT BIOQUEST
New Product
:
Nuclear Blue™ LCS1
AATB17559
:
AAT BIOQUEST
New Product
:
Pk:
0,5 mL
Seq:
2040
:
:
822.69 g/mol