Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
FMOC-Lys(QSY-7)-OH
FMOC-Lys(QSY-7)-OH
FMOC-Lys(QSY-7)-OH
FMOC-Lys(QSY-7)-OH
Catalog # AATB5261
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
FMOC-Lys(QSY-7)-OH
Catalog # AATB5261
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
50 mg
Seq:
2370
Specifications
About this item
Product Details
:
:
1008.2 g/mol
Product Details