Order Entry
Italy
ContactUsLinkComponent
Recombinant mcKLH (from Megathura crenulata)
Recombinant mcKLH (from Megathura crenulata)
Catalog # AATB5607
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Recombinant mcKLH (from Megathura crenulata)
Catalog # AATB5607
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Megathura crenulata
  • Storage conditions:
    Freeze (<−15 °C); Minimise light exposure
  • Protein synonyms:
    Mariculture keyhole limpet hemocyanin
  • Protein/peptide name:
    mcKLH
  • Shipping temperature:
    RT
  • Pk Info:
    10 mg

Specifications

About this item

Mariculture keyhole limpet hemocyanin (mcKLH) is a purified hemocyanin carrier protein optimised for producing highly effective immunogens with peptide antigens.

  • Designed for heightened immunogen response

mcKLH is compatible with hapten-carrier conjugation protocols in both phosphate and MES buffer systems, optimised for NHS-ester and EDC crosslinking methods, respectively.

Caution: For research use only.