Order Entry
Italy
Orders LinkContactUsLinkComponent
Ammonium High Calibration Standard

Ammonium High Calibration Standard

Ammonium high calibration standard, HDPE can, colourless, 1 L
Catalog # PRLS90344.290
Supplier:   PROLABO CMP
Ammonium High Calibration Standard
Catalog # PRLS90344.290
Supplier:  PROLABO CMP
CAS Number:  
Ammonium High Calibration Standard

Ammonium high calibration standard, HDPE can, colourless, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents

In accordance with the operating mode: 4 ml Ammonium stock solution 1000 ppm QSP 1000 ml Brine solution (320 g Sodium Chloride Ref Prolabo 27810, QSP 1000 ml deionised water).

Specifications

Specifications

  • Pack type:
    HDPE can, colourless
  • Pk:
    1 L
  • Description:
    Ammonium high calibration standard

Specifications