Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AzoDye-3 carboxylic acid
AzoDye-3 carboxylic acid
AzoDye-3 carboxylic acid
AzoDye-3 carboxylic acid
Catalog # AATB2470
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
AzoDye-3 carboxylic acid
Catalog # AATB2470
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
Storage Temperature:
Freezer
:
583.13 g/mol
Product Details