Order Entry
Italy
ContactUsLinkComponent
Human Recombinant AFT2 (from sf9 cells)
Human Recombinant AFT2 (from sf9 cells)
Catalog # ORIGTP710017
Supplier:  OriGene
New Product
CAS Number:  
Human Recombinant AFT2 (from sf9 cells)
Catalog # ORIGTP710017
Supplier:  OriGene
New Product
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Sf9 cells
  • Species:
    Human
  • Tag sequence:
    C-terminal polyhistidine tag
  • Storage conditions:
    Store at –80 °C
  • Biological activity:
    In vitro kinase assay substrate
  • Protein synonyms:
    TREB7|HB16|CREB2|CREB-2|CRE-BP1
  • UniProtKB:
    P15336
  • Protein/peptide name:
    ATF2
  • Purity:
    >80% as determined by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    55 kDa
  • Sequence:
    A DNA sequence from TrueORF clone, RC218983, encoding human full-length ATF2
  • Concentration:
    >0,05 µg/µl as determined by microplate BCA method

Specifications

About this item

Recombinant protein of human activating transcription factor 2 (ATF2), full length, with C-terminal polyhistidine tag, expressed in sf9 cells.

Buffer: 50 mM Tris-HCl, pH 8,0, 150 mM NaCl, 20% glycerol