Haematoxylin
Catalog # MHS80-2.5L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Appearance (Colour):Red-Brown
- Appearance (Form):Solution
- Appearance (Turbidity):Clear
- Performance:Conforms
- Expiration Date Period:36 Months
Specification Test Results
About this item
:
:302.28 g/mol
:M-Clarity™ quality level = MQ500 (more info.)
CAS Number:517-28-2
:208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111