Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:55 kDa erythrocyte membrane protein
- Simbolo dell'antigene:MPP1
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Citometria a flusso:Yes
- Ospite:Rabbit
- ImmunoChimica:Yes
- Reattività:Human
- Western blot:Yes
- Epitopo:Amino Acid Range = 301-327
- Forma:Liquid
- ID gene:Q00013
- Sinomini antigene:MPP1
- Modifica:None
- Tampone di conservazione:Supplied in PBS with 0,09% (W/V) sodium azide
- Peso molecolare:52 kDa
- Temperatura di conservazione:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentrazione:0,5 mg/ml
- Temperatura di spedizione:4 °C
- Immunogeno:This MPP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 301-327 amino acids from the Central region of human MPP1.
- Purificazione:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Taglia:400 µl
- Conf:400 µl
Specifications
About this item
This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: MPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human