Order Entry
Italy
ContactUsLinkComponent
Anti-MOPC21 Recombinant Antibody [Clone: MOPC-21]
Anti-MOPC21 Recombinant Antibody [Clone: MOPC-21]
Catalog # PRSI24-215
Supplier:  ProSci Inc.
Anti-MOPC21 Recombinant Antibody [Clone: MOPC-21]
Catalog # PRSI24-215
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Clone:
    MOPC-21
  • Conjugation:
    Unconjugated
  • Isotype:
    Fab fragment kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    MOPC21
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Tested applications:
    Negative control
  • Size:
    0,2 mg
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: MOPC21
Clonality: Recombinant
Clone: MOPC-21
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: