Order Entry
Italy
ContactUsLinkComponent
Anti-tag-72 Mouse Monoclonal Antibody [clone: TAGP-4]
Anti-tag-72 Mouse Monoclonal Antibody [clone: TAGP-4]
Catalog # PRSI33-907
Supplier:  ProSci Inc.
Anti-tag-72 Mouse Monoclonal Antibody [clone: TAGP-4]
Catalog # PRSI33-907
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    mRNA cap guanine-N7 methyltransferase
  • Antigen symbol:
    tag-72
  • Clonality:
    Monoclonal
  • Clone:
    TAGP-4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    Q9XVS1
  • Antigen synonyms:
    tag-72
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Recombinant human TAG-72 protein was used as the immunogen for this antibody.
  • Purification:
    Protein G affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This antibody recognises an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of TAG-72 antibody with a GCDFP-15 antibody is valuable in the diagnosis of apocrine carcinoma.

Flow Cytometry: 0,5-1 µg/million cells IF: 1 - 2 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the TAG-72 antibody to be titered up or down for optimal performance.

1. No pretreatment is required for staining of formalin-fixed, paraffin-embedded tissues.

Type: Primary
Antigen: tag-72
Clonality: Monoclonal
Clone: TAGP-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human