Order Entry
Italy
ContactUsLinkComponent
Anti-KRT76 Rabbit Monoclonal Antibody [clone: KRTH/1576R]
Anti-KRT76 Rabbit Monoclonal Antibody [clone: KRTH/1576R]
Catalog # PRSI33-833
Supplier:  ProSci Inc.
Anti-KRT76 Rabbit Monoclonal Antibody [clone: KRTH/1576R]
Catalog # PRSI33-833
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 76
  • Antigen symbol:
    KRT76
  • Clonality:
    Monoclonal
  • Clone:
    KRTH/1576R
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    Q01546
  • Antigen synonyms:
    KRT76
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Recombinant human KRT76 protein was used as the immunogen for the recombinant Basic Cytokeratin antibody.
  • Purification:
    Protein A affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This mAb recognises basic (Type II or HMW) cytokeratins, which include 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8). Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50 , 48, 46, 45, and 40 kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,25-0,5 µg/ml for 30 min at RT Prediluted IHC only format: incubate for 30 min at RT (1) Optimal dilution of the recombinant Basic Cytokeratin antibody should be determined by the researcher.

1. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: KRT76
Clonality: Monoclonal
Clone: KRTH/1576R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG, k
Reactivity: Human