Specifications
- Antibody type:Primary
- Antigen name:Tumor-associated Glycoprotein
- Antigen symbol:TAG-72 Antibody [CC49]
- Clonality:Monoclonal
- Clone:CC49
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine,Dog,Rat
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Purified TAG-72 protein was used as the immunogen for this TAG-72 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recognises an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This mAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma.
Western blot: 1 - 2 µg/ml Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the TAG-72 antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: TAG-72 Antibody [CC49]
Clonality: Monoclonal
Clone: CC49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Dog, Rat