Specifiche
- Antibody type:Primary
- Antigen name:Histo-blood group ABO system transferase
- Antigen symbol:ABO
- Clonality:Monoclonal
- Clone:HEB-29
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Neutralisation:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P16442
- Antigen synonyms:ABO
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:A mixture of erythrocytes of group B and glycoprotein fraction isolated from saliva of secretors with blood group B was used as the immunogen for the ABO antibody.
- Tested applications:Functional
- Purification:PEG precipitation
- Size:100 µg
- Pk:100 µG
Specifiche
Informazioni su questo articolo
The antibody HEB-29 reacts with human blood group B. The specificity of the antibody HEB-29 was confirmed by comparison of specificity and reactivity to standard reagent using >5,000 samples of blood. mAb HEB-29 shows specific staining of erythrocytes and vascular epithelium of blood group B controls and no staining in group A controls. It is applicable for tissue staining in tumor patients with blood groups B and AB. Blood group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Functional Activity-Erythrocyte Agglutination Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Optimal dilution of the ABO antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
Type: Primary
Antigen: ABO
Clonality: Monoclonal
Clone: HEB-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human