Specifications
- Antibody type:Primary
- Antigen name:glycoprotein hormones, alpha polypeptide
- Antigen symbol:CG alpha
- Clonality:Monoclonal
- Clone:HCGa/53
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P01215
- Antigen synonyms:CG alpha
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant protein was used as the immunogen for this HCG-alpha antibody.
- Purification:Protein G purified monoclonal antibody
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This antibody reacts with a protein of ~13 kDa, identified as alpha sub-unit of HCG. HCG is a glycoprotein which is secreted in large quantities by normal trophoblasts. HCG is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).
IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) Variations in protocols, secondaries and substrates may require the HCG-alpha antibody to be titered for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CG alpha
Clonality: Monoclonal
Clone: HCGa/53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human