Anti-SLC17A6 Rabbit Polyclonal Antibody
Catalog # PRSI15-915
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 17 (vesicular glutamate transporter), member 6
- Antigen symbol:SLC17A6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:57084
- Antigen synonyms:member 6|solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter)|differentiation-associated Na(+)-dependent inorganic phosphate cotransporter|differentiation-associated BNPI|SLC17A6|differentiation-associated Na-de|DNPI|VGLUT2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human SLC17A6 (NP_065079.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC17A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat