Anti-PLXNA3 Rabbit Polyclonal Antibody
Catalog # PRSI15-786
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Plexin A3
- Antigen symbol:PLXNA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:55558
- Antigen synonyms:XAP-6|Sex chromosome X transmembrane protein of HGF receptor family 3|plexin-4|PLXN4|PLXN3|HSSEXGENE|6.3|semaphorin receptor SEX|plexin-A3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 950-1150 of human PLXNA3 (NP_059984.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PLXNA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse